Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGT76E2
Entrez Gene ID
836078
Gene Name
UDP-glucosyl transferase 76E2
Gene Aliases
AT5G59590, F2O15.19, F2O15_19, UDP-glucosyl transferase 76E2
Chromosome Location
Chr.5: 24008865 - 24010688 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125351.3 NP_200767.1 1 - 2 587 67
GenBank mRNA BT004159.1 - 1 - 2 548 67
BT005494.1 - 1 - 2 484 67
KJ138632.1 - 1 - 2 484 67

View Full Product Details