Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GH9B10
Entrez Gene ID
819063
Gene Name
glycosyl hydrolase 9B10
Gene Aliases
AT2G44550, AtGH9B10, F4I1.55, glycosyl hydrolase 9B10
Chromosome Location
Chr.2: 18389381 - 18391142 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130019.1 NP_181983.1 1 - 2 227 139

View Full Product Details