Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DBE1
Entrez Gene ID
839531
Gene Name
debranching enzyme 1
Gene Aliases
ARABIDOPSIS THALIANA ISOAMYLASE 2, AT1G03310, ATISA2, AtBE2, BE2, BRANCHING ENZYME 2, F15K9.9, F15K9_9, ISA2, debranching enzyme 1
Chromosome Location
Chr.1: 813413 - 816812 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100213.4 NP_171830.1 2 - 2 2819 79
NM_202022.3 NP_973751.1 2 - 2 2842 79

View Full Product Details