Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBQ4
Entrez Gene ID
832184
Gene Name
ubiquitin 4
Gene Aliases
AT5G20620, T1M15.20, T1M15_20, ubiquitin 4
Chromosome Location
Chr.5: 6973053 - 6975009 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343676.1 NP_001332370.1 3 - 3 1152 135
NM_122069.4 NP_568397.1 2 - 2 1333 135

View Full Product Details