Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELF4-L4
Entrez Gene ID
838318
Gene Name
ELF4-like 4
Gene Aliases
AT1G17455, ELF4-like 4
Chromosome Location
Chr.1: 5997057 - 5998599 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160876.1 NP_001154348.1 2 - 2 632 137
NM_101607.4 NP_564024.1 2 - 2 763 137
GenBank mRNA AK220611.1 - 2 - 2 556 137

View Full Product Details