Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AVA-P2
Entrez Gene ID
838579
Gene Name
ATPase, F0/V0 complex, subunit C protein
Gene Aliases
AT1G19910, ATVHA-C2, AVA-2PE, F6F9.3, F6F9_3, VACUOLAR H+-PUMPING ATPASE 16 KDA PROTEOLIPID, VACUOLAR-TYPE H+ ATPASE C2
Chromosome Location
Chr.1: 6913001 - 6914634 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332418.1 NP_001321379.1 3 - 3 1029 94
NM_101846.4 NP_564098.2 3 - 3 801 94
GenBank mRNA L44585.1 - 3 - 3 493 94

View Full Product Details