Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPK7
Entrez Gene ID
816330
Gene Name
MAP kinase 7
Gene Aliases
AT2G18170, ATMPK7, F8D23.5, F8D23_5, MAP kinase 7
Chromosome Location
Chr.2: 7907774 - 7910040 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127374.4 NP_179409.1 3 - 3 1743 149

View Full Product Details