Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CZF1
Entrez Gene ID
818605
Gene Name
zinc finger (CCCH-type) family protein
Gene Aliases
(SALT-INDUCIBLE ZINC FINGER 2, AT2G40140, ATSZF2, SZF2, T7M7.3, T7M7_3, ZFAR1
Chromosome Location
Chr.2: 16771997 - 16774734 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036440.2 NP_001031517.1 2 - 2 1904 128
NM_129572.5 NP_181543.1 2 - 2 2114 128
GenBank mRNA AY093016.1 - 2 - 2 1951 128

View Full Product Details