Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS28-1
Entrez Gene ID
828241
Gene Name
vacuolar protein sorting-associated protein-like protein
Gene Aliases
AT4G21560, F17L22.20, VPS28, vacuolar protein sorting 28, vacuolar protein sorting-associated protein 28 homolog 1
Chromosome Location
Chr.4: 11468666 - 11470569 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341499.1 NP_001320024.1 2 - 2 1082 133
NM_118276.4 NP_193887.1 3 - 3 1186 133
NM_202856.3 NP_974585.1 2 - 2 1521 133
GenBank mRNA AY136475.1 - 3 - 3 1149 133

View Full Product Details