Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLP31
Entrez Gene ID
843421
Gene Name
MLP-like protein 31
Gene Aliases
AT1G70840, F15H11.9, F15H11_9, MLP-like protein 31
Chromosome Location
Chr.1: 26712982 - 26714069 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105752.4 NP_177241.3 2 - 2 632 129
GenBank mRNA AK226451.1 - 2 - 2 623 129
AY080814.1 - 2 - 2 628 129

View Full Product Details