Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR50
Entrez Gene ID
3768693
Gene Name
low-molecular-weight cysteine-rich 50
Gene Aliases
AT2G12465, low-molecular-weight cysteine-rich 50
Chromosome Location
Chr.2: 5055415 - 5055880 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036272.2 NP_001031349.1 1 - 2 92 119

View Full Product Details