Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR9
Entrez Gene ID
3768128
Gene Name
low-molecular-weight cysteine-rich 9
Gene Aliases
AT2G27145, low-molecular-weight cysteine-rich 9
Chromosome Location
Chr.2: 11600169 - 11600764 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036352.2 NP_001031429.1 1 - 2 124 91

View Full Product Details