Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR76
Entrez Gene ID
3767941
Gene Name
low-molecular-weight cysteine-rich 76
Gene Aliases
AT2G31953, low-molecular-weight cysteine-rich 76
Chromosome Location
Chr.2: 13583376 - 13584020 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036383.2 NP_001031460.1 1 - 2 112 139

View Full Product Details