Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCRL14
Entrez Gene ID
3770429
Gene Name
SCR-like 14
Gene Aliases
AT4G22115, SCR-like 14
Chromosome Location
Chr.4: 11714743 - 11715243 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036617.2 NP_001031694.1 1 - 2 148 69

View Full Product Details