Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR19
Entrez Gene ID
3770561
Gene Name
low-molecular-weight cysteine-rich 19
Gene Aliases
AT4G30074, low-molecular-weight cysteine-rich 19
Chromosome Location
Chr.4: 14699737 - 14700599 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036678.2 NP_001031755.1 1 - 2 95 82

View Full Product Details