Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NdhM
Entrez Gene ID
3770591
Gene Name
subunit NDH-M of NAD(P)H:plastoquinone dehydrogenase complex
Gene Aliases
AT4G37925, NADH dehydrogenase-like complex M, NDH-M, subunit NDH-M of NAD(P)H:plastoquinone dehydrogenase complex
Chromosome Location
Chr.4: 17830533 - 17831619 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036727.3 NP_001031804.1 1 - 2 588 108

View Full Product Details