Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- VIM4
- Entrez Gene ID
-
842917
- Gene Name
- Zinc finger (C3HC4-type RING finger) family protein
- Gene Aliases
-
AT1G66040, F15E12.8, F15E12_8, ORTH4, ORTHRUS 4, VARIANT IN METHYLATION 4
- Chromosome Location
-
Chr.1: 24583740 - 24586760 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105275.2 |
NP_176778.1 |
1 - 1 |
221 |
130 |
View Full Product Details