Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRSF6
Entrez Gene ID
507828
Gene Name
serine and arginine rich splicing factor 6
Gene Aliases
SFRS6
Chromosome Location
Chr.13: 72096697 - 72102392 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035272.1 NP_001030349.1 2 - 3 399 59
GenBank mRNA BC107553.1 - 2 - 3 399 59

View Full Product Details