Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GATAD2B
Entrez Gene ID
505039
Gene Name
GATA zinc finger domain containing 2B
Gene Aliases
-
Chromosome Location
Chr.3: 16506949 - 16590722 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192047.1 NP_001178976.1 7 - 8 1419 83

View Full Product Details