Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGA5
Entrez Gene ID
414350
Gene Name
pepsinogen 5, group I (pepsinogen A)
Gene Aliases
pga
Chromosome Location
Chr.29: 39760936 - 39771371 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001600.2 NP_001001600.2 6 - 7 802 66
GenBank mRNA AY330769.1 - 5 - 6 723 66
BC149644.1 - 5 - 6 752 66

View Full Product Details