Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP20
Entrez Gene ID
445424
Gene Name
cilia and flagella associated protein 20
Gene Aliases
C16orf80, C18H16orf80, GTL3
Chromosome Location
Chr.18: 25994997 - 26008254 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003905.1 NP_001003905.1 1 - 2 320 114
GenBank mRNA AY675080.1 - 1 - 2 320 114
BC112831.1 - 1 - 2 308 114

View Full Product Details