Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFAIP6
Entrez Gene ID
493710
Gene Name
TNF alpha induced protein 6
Gene Aliases
tsg-6
Chromosome Location
Chr.2: 44746822 - 44763774 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007813.2 NP_001007814.1 1 - 2 161 154
GenBank mRNA AJ854251.1 - 1 - 2 163 154
BC151789.1 - 1 - 2 175 154

View Full Product Details