Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC2A12
Entrez Gene ID
497031
Gene Name
solute carrier family 2 member 12
Gene Aliases
GLUT12
Chromosome Location
Chr.9: 72148823 - 72225025 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011683.2 NP_001011683.1 2 - 3 1546 60
GenBank mRNA AY514443.1 - 2 - 3 1547 60

View Full Product Details