Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BREH1
Entrez Gene ID
497207
Gene Name
retinyl ester hydrolase type 1
Gene Aliases
-
Chromosome Location
Chr.18: 24745125 - 24771961 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012287.1 NP_001012287.1 10 - 11 1164 93
GenBank mRNA AY369075.1 - 10 - 11 1164 93
BC102781.1 - 10 - 11 1215 93

View Full Product Details