Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TM4SF4
Entrez Gene ID
507646
Gene Name
transmembrane 4 L six family member 4
Gene Aliases
-
Chromosome Location
Chr.1: 118663092 - 118690685 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034306.2 NP_001029478.1 4 - 5 777 67
GenBank mRNA BC103284.1 - 4 - 5 778 67

View Full Product Details