Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCN1
Entrez Gene ID
508941
Gene Name
cellular communication network factor 1
Gene Aliases
CYR61, GIG1
Chromosome Location
Chr.3: 58488393 - 58491302 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034340.2 NP_001029512.1 4 - 5 1069 97
GenBank mRNA BC102934.1 - 4 - 5 1070 97

View Full Product Details