Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZSCAN21
Entrez Gene ID
527103
Gene Name
zinc finger and SCAN domain containing 21
Gene Aliases
ZNF38
Chromosome Location
Chr.25: 36387591 - 36395341 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035368.2 NP_001030445.1 2 - 3 520 75
GenBank mRNA BC103036.1 - 2 - 3 521 75

View Full Product Details