Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFS7
Entrez Gene ID
338079
Gene Name
NADH:ubiquinone oxidoreductase core subunit S7
Gene Aliases
-
Chromosome Location
Chr.7: 43807778 - 43814268 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038022.1 NP_001033111.1 4 - 5 253 63
GenBank mRNA BC109716.1 - 4 - 5 258 63
BC153837.1 - 3 - 4 660 63
X65020.1 - - 253 63

View Full Product Details