Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C21H15orf40
Entrez Gene ID
509050
Gene Name
chromosome 21 C15orf40 homolog
Gene Aliases
C15orf40
Chromosome Location
Chr.21: 23257778 - 23264117 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075386.2 NP_001068854.1 1 - 2 82 65
GenBank mRNA BC103179.1 - 1 - 2 84 65

View Full Product Details