Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UQCR11
Entrez Gene ID
281570
Gene Name
ubiquinol-cytochrome c reductase, complex III subunit XI
Gene Aliases
UQCR
Chromosome Location
Chr.7: 43976258 - 43979178 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174215.2 NP_776640.1 1 - 2 63 97
GenBank mRNA BC102080.1 - 1 - 2 89 97
BC126774.1 - 1 - 2 67 97
D55637.1 - 1 - 2 63 97

View Full Product Details