Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CATHL5
Entrez Gene ID
282167
Gene Name
cathelicidin 5
Gene Aliases
-
Chromosome Location
Chr.22: 51671494 - 51674373 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174510.3 NP_776935.1 3 - 4 400 79
GenBank mRNA BC142015.1 - 3 - 4 404 79
X97609.1 - - 402 79

View Full Product Details