Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYBB1
Entrez Gene ID
282205
Gene Name
crystallin beta B1
Gene Aliases
-
Chromosome Location
Chr.17: 66251958 - 66265139 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174526.2 NP_776951.1 1 - 2 206 75
GenBank mRNA X01808.1 - - 206 75

View Full Product Details