Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCGR2B
Entrez Gene ID
282229
Gene Name
Fc fragment of IgG receptor IIb
Gene Aliases
CD32, FCGR2, fcRII
Chromosome Location
Chr.3: 7931199 - 7947831 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174539.2 NP_776964.1 2 - 3 206 59
GenBank mRNA BC113215.1 - 2 - 3 194 59
EU151550.1 - 2 - 3 127 59
X75671.1 - - 206 59

View Full Product Details