Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP3R1
Entrez Gene ID
282321
Gene Name
protein phosphatase 3 regulatory subunit B, alpha
Gene Aliases
-
Chromosome Location
Chr.11: 66605841 - 66667440 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174583.2 NP_777008.1 4 - 5 340 98
GenBank mRNA AB375794.1 - 2 - 3 229 98
X71666.1 - - 340 98

View Full Product Details