Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STX1B
Entrez Gene ID
282377
Gene Name
syntaxin 1B
Gene Aliases
STX1B2
Chromosome Location
Chr.25: 27123000 - 27158868 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174618.2 NP_777043.1 9 - 10 836 79
GenBank mRNA BC150124.1 - 9 - 10 786 79
D14133.1 - 9 - 10 836 79

View Full Product Details