Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPK3A
Entrez Gene ID
100336102
Gene Name
uroplakin 3A
Gene Aliases
UP3a, UPIII, Upk3
Chromosome Location
Chr.5: 115685872 - 115706832 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174709.3 NP_777134.1 3 - 4 521 72

View Full Product Details