Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNA3
Entrez Gene ID
282702
Gene Name
cholinergic receptor nicotinic alpha 3 subunit
Gene Aliases
-
Chromosome Location
Chr.5: 71462099 - 71462455 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174719.2 NP_777144.1 3 - 4 598 90
GenBank mRNA BC142063.1 - 3 - 4 275 90
X57032.1 - - 598 90

View Full Product Details