Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYL6
Entrez Gene ID
281341
Gene Name
myosin light chain 6
Gene Aliases
-
Chromosome Location
Chr.5: 57161868 - 57165070 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175780.1 NP_786974.1 1 - 2 33 57
GenBank mRNA BC103428.1 - 1 - 2 47 57
X54978.1 - - 33 57

View Full Product Details