Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SULT1E1
Entrez Gene ID
280934
Gene Name
sulfotransferase family 1E member 1
Gene Aliases
ST1E1, STE
Chromosome Location
Chr.6: 85309022 - 85365114 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177488.3 NP_803454.2 8 - 9 995 86
GenBank mRNA BC102939.1 - 7 - 8 895 86
EV793129.1 - 5 - 6 570 86
M54942.1 - 9 - 10 991 86
X56395.1 - - 991 86

View Full Product Details