Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TREM1
Entrez Gene ID
404547
Gene Name
triggering receptor expressed on myeloid cells 1
Gene Aliases
TREM-1
Chromosome Location
Chr.23: 15140492 - 15155609 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206970.1 NP_996853.1 2 - 3 418 90
GenBank mRNA AY525122.1 - 2 - 3 418 90

View Full Product Details