Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C15H11orf16
Entrez Gene ID
504224
Gene Name
chromosome 15 C11orf16 homolog
Gene Aliases
C11orf16
Chromosome Location
Chr.15: 43659755 - 43669711 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014846.1 NP_001014846.1 3 - 4 506 157
GenBank mRNA BC102543.1 - 4 - 5 577 157
BT021678.1 - 3 - 4 506 157

View Full Product Details