Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PRICKLE3
- Entrez Gene ID
-
509780
- Gene Name
- prickle planar cell polarity protein 3
- Gene Aliases
-
LMO6
- Chromosome Location
-
Chr.X: 87219384 - 87228549 on Build ARS-UCD1.2
- Gene Symbol
- BOLA
- Entrez Gene ID
-
515712
- Gene Name
- MHC class I heavy chain
- Gene Aliases
-
3349.1, BoLA-N, Bota, D18.5, LA-A11
- Chromosome Location
-
Chr.23: 28720499 - 28724353 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC149733.1 |
- |
- |
103 |
136 |
View Full Product Details