Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAJB5
Entrez Gene ID
540083
Gene Name
DnaJ heat shock protein family (Hsp40) member B5
Gene Aliases
-
Chromosome Location
Chr.8: 59277545 - 59286524 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014959.1 NP_001014959.1 3 - 4 954 74
GenBank mRNA BC120323.1 - 3 - 4 914 74
BT021176.1 - 3 - 4 954 74

View Full Product Details