Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF2B2
Entrez Gene ID
513958
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Gene Aliases
-
Chromosome Location
Chr.10: 86259480 - 86266557 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015593.1 NP_001015593.1 2 - 3 310 97
GenBank mRNA BC123823.1 - 2 - 3 314 97
BT021006.1 - 2 - 3 310 97

View Full Product Details