Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARMC1
Entrez Gene ID
514000
Gene Name
armadillo repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.14: 30000050 - 30024834 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015594.3 NP_001015594.2 6 - 7 888 85
GenBank mRNA BC103407.1 - 6 - 7 889 85
BT020641.1 - 6 - 7 839 85
EV727326.1 - 1 - 2 470 85

View Full Product Details