Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DHODH
Entrez Gene ID
533873
Gene Name
dihydroorotate dehydrogenase (quinone)
Gene Aliases
-
Chromosome Location
Chr.18: 39068792 - 39081933 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015650.1 NP_001015650.1 7 - 8 977 63
GenBank mRNA BC120337.1 - 7 - 8 977 63
BT020807.1 - 7 - 8 977 63
BT026309.1 - 7 - 8 966 63

View Full Product Details