Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLR3D
Entrez Gene ID
508049
Gene Name
RNA polymerase III subunit D
Gene Aliases
-
Chromosome Location
Chr.8: 69558203 - 69564549 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024499.1 NP_001019670.1 1 - 2 87 64
GenBank mRNA BT021699.1 - 1 - 2 87 64

View Full Product Details