Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRP9
Entrez Gene ID
520716
Gene Name
NLR family pyrin domain containing 9
Gene Aliases
NALP9
Chromosome Location
Chr.18: 61771669 - 61796493 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024664.1 NP_001019835.1 3 - 4 2007 115
GenBank mRNA DQ011673.1 - 3 - 4 2007 115
DQ092866.1 - 3 - 4 115

View Full Product Details