Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBP1
Entrez Gene ID
537379
Gene Name
retinol binding protein 1
Gene Aliases
CRBP1
Chromosome Location
Chr.1: 129556754 - 129583703 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025343.1 NP_001020514.1 3 - 4 482 104
GenBank mRNA AF502256.1 - 3 - 4 482 104
BC109589.1 - 3 - 4 401 104
BT021637.1 - 7 - 8 2503 104

View Full Product Details