Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NANOG
Entrez Gene ID
538951
Gene Name
Nanog homeobox
Gene Aliases
-
Chromosome Location
Chr.5: 101412795 - 101417948 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025344.1 NP_001020515.1 1 - 2 317 92
GenBank mRNA AM039957.1 - 1 - 2 80 92
DQ069776.1 - 1 - 2 317 92

View Full Product Details